SY3H5815F785 Synthonix Corporation

2,4-Dibromo-1-nitrobenzene

  • Cas Number : 51686-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24039.html

Safety Information

MSDS https://synthonix.com/24039.html
Availability details for
Quantity
  • Availability
  • Add to Cart