SY3H5815F786 Synthonix Corporation

3,4-Dihydro-2H-pyrano[2,3-c]pyridine-6-carbaldehyde

  • Cas Number : 527681-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24093.html

Safety Information

MSDS https://synthonix.com/24093.html
Availability details for
Quantity
  • Availability
  • Add to Cart