SY3H5815F787 Synthonix Corporation

(4,6-Dimethylpyrimidin-2-yl)methanol

  • Cas Number : 54198-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24164.html

Safety Information

MSDS https://synthonix.com/24164.html
Availability details for
Quantity
  • Availability
  • Add to Cart