SY3H5815F78F Synthonix Corporation

2,6-Dichloro-3-methyl-5-nitropyridine

  • Cas Number : 58596-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24400.html

Safety Information

MSDS https://synthonix.com/24400.html
Availability details for
Quantity
  • Availability
  • Add to Cart