SY3H5815F790 Synthonix Corporation

3,3-Dimethylmorpholine

  • Cas Number : 59229-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24429.html

Safety Information

MSDS https://synthonix.com/24429.html
Availability details for
Quantity
  • Availability
  • Add to Cart