SY3H5815F79F Synthonix Corporation

2,5-Dichloro-N-(2-(isopropylsulfonyl)phenyl)pyrimidin-4-amine

  • Cas Number : 761440-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25269.html

Safety Information

MSDS https://synthonix.com/25269.html
Availability details for
Quantity
  • Availability
  • Add to Cart