SY3H5815F7A0 Synthonix Corporation

5,6-Dimethoxy-1H-indazole

  • Cas Number : 7746-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25346.html

Safety Information

MSDS https://synthonix.com/25346.html
Availability details for
Quantity
  • Availability
  • Add to Cart