SY3H5815F7A7 Synthonix Corporation

4,6-Dichloro-1-methyl-1H-imidazo[4,5-c]pyridine

  • Cas Number : 887147-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/26193.html

Safety Information

MSDS https://synthonix.com/26193.html
Availability details for
Quantity
  • Availability
  • Add to Cart