SY3H5815F7B9 Synthonix Corporation

(3R,5R)-3,5-Dimethylmorpholine hydrochloride

  • Cas Number : 1542268-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/27896.html

Safety Information

MSDS https://synthonix.com/27896.html
Availability details for
Quantity
  • Availability
  • Add to Cart