SY3H5815F7C2 Synthonix Corporation

4,6-Dimethoxypyrimidine

  • Cas Number : 5270-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3513.html

Safety Information

MSDS https://synthonix.com/3513.html
Availability details for
Quantity
  • Availability
  • Add to Cart