SY3H5815F7CB Synthonix Corporation

2,4-Diamino-6-methoxypyrimidine

  • Cas Number : 3270-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3978.html

Safety Information

MSDS https://synthonix.com/3978.html
Availability details for
Quantity
  • Availability
  • Add to Cart