SY3H5815F7F1 Synthonix Corporation

(1-Ethyl-1H-pyrazol-5-yl)boronic acid

  • Cas Number : 1095080-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16540.html

Safety Information

MSDS https://synthonix.com/16540.html
Availability details for
Quantity
  • Availability
  • Add to Cart