SY3H5815F7F3 Synthonix Corporation

4-(4-Ethylpiperazin-1-yl)phenylamine

  • Cas Number : 115619-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16821.html

Safety Information

MSDS https://synthonix.com/16821.html
Availability details for
Quantity
  • Availability
  • Add to Cart