SY3H5815F7F6 Synthonix Corporation

(R)-Ethyl piperidine-3-carboxylate hydrochloride

  • Cas Number : 37675-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1816.html

Safety Information

MSDS https://synthonix.com/1816.html
Availability details for
Quantity
  • Availability
  • Add to Cart