SY3H5815F7FD Synthonix Corporation

Ethyl 6-hydroxy-2-naphthoate

  • Cas Number : 17295-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/21806.html

Safety Information

MSDS https://synthonix.com/21806.html
Availability details for
Quantity
  • Availability
  • Add to Cart