SY3H5815F7FF Synthonix Corporation

Ethyl 2-(4-bromophenyl)-2-oxoacetate

  • Cas Number : 20201-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22198.html

Safety Information

MSDS https://synthonix.com/22198.html
Availability details for
Quantity
  • Availability
  • Add to Cart