SY3H5815F806 Synthonix Corporation

1-(4-Ethylphenyl)propan-1-one

  • Cas Number : 27465-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22775.html

Safety Information

MSDS https://synthonix.com/22775.html
Availability details for
Quantity
  • Availability
  • Add to Cart