SY3H5815F807 Synthonix Corporation

(R)-Ethyl 3-methylpiperidine-3-carboxylate

  • Cas Number : 297172-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22903.html

Safety Information

MSDS https://synthonix.com/22903.html
Availability details for
Quantity
  • Availability
  • Add to Cart