SY3H5815F808 Synthonix Corporation

Ethyl 5-nitropicolinate

  • MDL number MFCD09751108
  • Cas Number : 30563-98-5
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Yellow solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/22949.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/22949.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart