SY3H5815F809 Synthonix Corporation

Ethyl oxazole-2-carboxylate

  • Cas Number : 33036-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23077.html

Safety Information

MSDS https://synthonix.com/23077.html
Availability details for
Quantity
  • Availability
  • Add to Cart