SY3H5815F80A Synthonix Corporation

Ethyl 4-((3-chloro-4-methoxybenzyl)amino)-2-(methylthio)pyrimidine-5-carboxylate

  • Cas Number : 330785-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23081.html

Safety Information

MSDS https://synthonix.com/23081.html
Availability details for
Quantity
  • Availability
  • Add to Cart