SY3H5815F817 Synthonix Corporation

Ethyl 2-aminopyrimidine-5-carboxylate

  • Cas Number : 57401-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24339.html

Safety Information

MSDS https://synthonix.com/24339.html
Availability details for
Quantity
  • Availability
  • Add to Cart