SY3H5815F81A Synthonix Corporation

Ethyl 3-Methylpyridine-2-carboxylate

  • Cas Number : 58997-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24419.html

Safety Information

MSDS https://synthonix.com/24419.html
Availability details for
Quantity
  • Availability
  • Add to Cart