SY3H5815F844 Synthonix Corporation

7-Ethoxy-2-naphthalenol

  • Cas Number : 57944-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4952.html

Safety Information

MSDS https://synthonix.com/4952.html
Availability details for
Quantity
  • Availability
  • Add to Cart