SY3H5815F860 Synthonix Corporation

Ethyl 1-ethyl-3-methyl-1H-pyrazole-5-carboxylate

  • Cas Number : 50920-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/9935.html

Safety Information

MSDS https://synthonix.com/9935.html
Availability details for
Quantity
  • Availability
  • Add to Cart