SY3H5815F867 Synthonix Corporation

N-(5-Fluoro-2-oxo-2,3-dihydropyrimidin-4-yl)benzamide

  • Cas Number : 10357-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16183.html

Safety Information

MSDS https://synthonix.com/16183.html
Availability details for
Quantity
  • Availability
  • Add to Cart