SY3H5815F86A Synthonix Corporation

6-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine

  • Cas Number : 1190319-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17196.html

Safety Information

MSDS https://synthonix.com/17196.html
Availability details for
Quantity
  • Availability
  • Add to Cart