SY3H5815F888 Synthonix Corporation

5-Fluoroquinolin-8-ol

  • Cas Number : 387-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/23436.html

Safety Information

MSDS https://synthonix.com/23436.html
Availability details for
Quantity
  • Availability
  • Add to Cart