SY3H5815F891 Synthonix Corporation

5-Fluoro-2-methoxypyridin-4-amine

  • Cas Number : 58381-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/24390.html

Safety Information

MSDS https://synthonix.com/24390.html
Availability details for
Quantity
  • Availability
  • Add to Cart