SY3H5815F898 Synthonix Corporation

6-Formylpicolinonitrile

  • Cas Number : 85148-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25689.html

Safety Information

MSDS https://synthonix.com/25689.html
Availability details for
Quantity
  • Availability
  • Add to Cart