SY3H5815F8B5 Synthonix Corporation

8-Fluorochroman-4-amine hydrochloride

  • Cas Number : 191608-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/5362.html

Safety Information

MSDS https://synthonix.com/5362.html
Availability details for
Quantity
  • Availability
  • Add to Cart