SY3H5815F8E3 Synthonix Corporation

(6-Hydroxynaphthalen-2-yl)boronic acid

  • Cas Number : 173194-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3730.html

Safety Information

MSDS https://synthonix.com/3730.html
Availability details for
Quantity
  • Availability
  • Add to Cart