SY3H5815F8E9 Synthonix Corporation

tert-Butyl 3-hydroxy-3-(trifluoromethyl)pyrrolidine-1-carboxylate

  • Cas Number : 644970-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/7117.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7117.html
hstatements H302 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart