SY3H5815F8EB Synthonix Corporation

(1H-indol-7-yl)boronic acid

  • Cas Number : 210889-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/2302.html

Safety Information

MSDS https://synthonix.com/2302.html
Availability details for
Quantity
  • Availability
  • Add to Cart