SY3H5815F8EE Synthonix Corporation

3-Iodo-6-methyl-1H-pyrrolo[2,3-b]pyridine

  • Cas Number : 1000340-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/15846.html

Safety Information

MSDS https://synthonix.com/15846.html
Availability details for
Quantity
  • Availability
  • Add to Cart