SY3H5815F8F7 Synthonix Corporation

5-Iodopyrimidine

  • Cas Number : 31462-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/22993.html

Safety Information

MSDS https://synthonix.com/22993.html
Availability details for
Quantity
  • Availability
  • Add to Cart