SY3H5815F901 Synthonix Corporation

4-Iodoisoxazole

  • Cas Number : 847490-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/25626.html

Safety Information

MSDS https://synthonix.com/25626.html
Availability details for
Quantity
  • Availability
  • Add to Cart