SY3H5815F907 Synthonix Corporation

Imidazo[1,2-b]pyridazine

  • Cas Number : 766-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/3654.html

Safety Information

MSDS https://synthonix.com/3654.html
Availability details for
Quantity
  • Availability
  • Add to Cart