SY3H5815F90E Synthonix Corporation

Indole-7-carboxylic acid

  • MDL number MFCD00210442
  • Cas Number : 1670-83-3
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Greyish yellow red
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/4695.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/4695.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart