SY3H5815F912 Synthonix Corporation

3-Iodo-6-methoxy-1H-indazole

  • Cas Number : 936138-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/6150.html

Safety Information

MSDS https://synthonix.com/6150.html
Availability details for
Quantity
  • Availability
  • Add to Cart