SY3H5815F926 Synthonix Corporation

Methyl 4-chloro-1H-pyrazole-3-carboxylate

  • Cas Number : 1005584-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/15947.html

Safety Information

MSDS https://synthonix.com/15947.html
Availability details for
Quantity
  • Availability
  • Add to Cart