SY3H5815F92B Synthonix Corporation

1-Methyl-1H-indol-5-amine

  • Cas Number : 102308-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16073.html

Safety Information

MSDS https://synthonix.com/16073.html
Availability details for
Quantity
  • Availability
  • Add to Cart