SY3H5815F935 Synthonix Corporation

5-Methyl-1H-indazol-6-ol

  • Cas Number : 1082042-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16451.html

Safety Information

MSDS https://synthonix.com/16451.html
Availability details for
Quantity
  • Availability
  • Add to Cart