SY3H5815F936 Synthonix Corporation

Methyl 1-methyl-1H-indole-3-carboxylate

  • Cas Number : 108438-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16466.html

Safety Information

MSDS https://synthonix.com/16466.html
Availability details for
Quantity
  • Availability
  • Add to Cart