SY3H5815F937 Synthonix Corporation

5-Methyl-3-nitropicolinonitrile

  • Cas Number : 1089330-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16492.html

Safety Information

MSDS https://synthonix.com/16492.html
Availability details for
Quantity
  • Availability
  • Add to Cart