SY3H5815F93A Synthonix Corporation

6-Methyl-2-vinylpyridine

  • Cas Number : 1122-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16640.html

Safety Information

MSDS https://synthonix.com/16640.html
Availability details for
Quantity
  • Availability
  • Add to Cart