SY3H5815F942 Synthonix Corporation

(R)-5-Methylmorpholin-3-one

  • Cas Number : 119844-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17373.html

Safety Information

MSDS https://synthonix.com/17373.html
Availability details for
Quantity
  • Availability
  • Add to Cart