SY3H5815F943 Synthonix Corporation

Methyl 5-(methylsulfonyl)picolinate

  • Cas Number : 1201326-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17386.html

Safety Information

MSDS https://synthonix.com/17386.html
Availability details for
Quantity
  • Availability
  • Add to Cart