SY3H5815F946 Synthonix Corporation

Methyl 3-(benzyloxy)-1-(2,3-dihydroxypropyl)-4-oxo-1,4-dihydropyridine-2-carboxylate

  • Cas Number : 1206102-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/17442.html

Safety Information

MSDS https://synthonix.com/17442.html
Availability details for
Quantity
  • Availability
  • Add to Cart