SY3H5815F94D Synthonix Corporation

Methyl 4-bromo-2-methyl-1H-indole-6-carboxylate

  • Cas Number : 1260383-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/18183.html

Safety Information

MSDS https://synthonix.com/18183.html
Availability details for
Quantity
  • Availability
  • Add to Cart